Electroporation Cuvettes, 0.4 cm, red

Catalog number:  P46050

Invitrogen™  Related applications: Transformation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

DNA can be introduced into bacterial, yeast, and mammalian cells by the process of electroporation. Invitrogen offers electroporation cuvettes for use with mammalian, bacterial or yeast cells with the following convenient features:

• Compatible with virtually all common electroporation devices
• Gamma-irradiated and individually packaged to ensure sterility
• Color-coded snap cap for easy identification of gap size
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Size: 50⁄bag
Shipping Condition: Room Temperature

Contents & storage

Each bag contains 50 individually wrapped, sterile cuvettes.

Documents

Manuals & protocols

Recommended products