Use ProLong® Live Antifade Reagent for brightest fluorescent images with live cells
Invitrogen™

Phen Green™ FL, Diacetate, cell permeant

Catalog number:  P6763

 Related applications: Cell Viability, Proliferation & Function

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The green-fluorescent heavy metal indicator Phen Green™ FL, diacetate can be used to detect a broad range of ions, including Cu2+, Cu+, Fe2+, Hg2+, Pb2+, Cd2+, Zn2+ and Ni2+.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Indicator Type: Cadmium Indicator, Copper Indicator, Iron Indicator, Lead Indicator, Mercury Indicator, Nickel Indicator, Zinc Indicator
Indicator Properties: Fluorescent Dye-Based
For Use With (Equipment): Fluorescence Microscope, Microplate Reader
Cell Permeability: Cell-Permeant
Detection Method: Fluorescent
Product Line: Phen Green™
Product Size: 1 mg
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products