ProtoArray™ Blocking Buffer Kit
Catalog number:  PA055
Invitrogen™  Related applications: Pharma & Biopharma | Protein Assays and Analysis | Target & Lead Identification & Validation
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
The Blocking Buffer Kit consists of reagents sufficient for preparing Blocking Buffer and Wash Buffer to process 10 ProtoArray® Human Protein Microarrays for ProtoArray® protein-protein interaction (PPI), immune response biomarker profiling (IRBP), and antibody specificity profiling (ASP) applications. The kit contains one bottle each of Blocking Buffer and 10X Synthetic Block, and one tube of dithiothreitol (DTT).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Compatible Products: ProtoArray® Human Protein Microarrays for antibody specificity profiling (ASP), ProtoArray® Human Protein Microarrays for immune response biomarker profiling (IRBP), ProtoArray® Human Protein Microarrays for protein-protein interaction (PPI)
Product Line: ProtoArray®
Product Size: 1 kit
Contents & storage
Store the ProtoArray® Blocking Buffer Kit at -20°.
Documents
Manuals & protocols