Gibco™

IL13 Recombinant Human Protein

Catalog number:  PHC0134

 Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant Human Interleukin 13 (IL13) is a bioactive protein intended for use in cell culture applications. IL13 down-regulates macrophage produced cytokines such as TNF-α, IL-1, IL-6, IL-8, and IL-12 in response to LPS challenge or IFN-γ. IL13 induces monocyte differentiation and also induces B cell differentiation and proliferation.

High purity—no interference from other proteins or contaminants
High biological activity—more results with less protein
Proven compatibility—Gibco® proteins bioassayed with Gibco® media

High purity recombinant bioactive protein
Recombinant Human Interleukin 13 is one of our more than 250 available Gibco® recombinant proteins. To help ensure Gibco® growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein.

High biological activity
In-house activity testing of Gibco® Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, antiviral, and protease assays. Recombinant Human Interleukin 13 (IL13) specific activity is determined by the dose dependent proliferation of human TF-1 cells.

Proven compatibility
Gibco® proteins are bioassayed with Gibco® media. Since 1962, Gibco® has been the consistent provider of high quality media, reagents, and sera for reliable cell culture.

See our selection of recombinant Gibco® growth factors.

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Immunology, Inflammation, Respiratory Research, Signal Transduction
Endotoxin Level: < 0.1 ng/µg
Expression: E. coli
Form: Lyophilized
Gene Aliases: IL-13
Gene ID: 3596
Mass: 5 µg
Molecular Weight: 17 kDa
Product Line: Gibco®
Product Size: 5 µg
Protein Family: Cytokines & Receptors
Protein Form: Ligand, Recombinant
Protein Subtype: Interleukins
Purification: Sequential Chromatography
Purity: 95 %
Purity or Quality Grade: Carrier-Free
Species: Human
Validated Application: Cell Culture
Shipping Condition: Approved for shipment on Wet or Dry Ice

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products