IL28A Recombinant Human Protein

Catalog number: PHC0284

Gibco™  Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant Human Interleukin-28A (IL28A) is a bioactive protein intended for use in cell culture applications. IL28A is involved in the immune response, with functionality similar to type I interferons. IL28A has been shown to bind a heterodimeric receptor comprised of IL-10 receptor beta and IL-28 receptor alpha.

• High purity—no interference from other proteins or contaminants
• High biological activity—more results with less protein
• Proven compatibility—Gibco® proteins bioassayed with Gibco® media

High purity recombinant bioactive protein
Recombinant Human Interleukin-28A (IL28A) is one of our more than 250 available Gibco® recombinant proteins. To help ensure Gibco® growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to get a biologically active protein.

High biological activity
In-house activity testing of Gibco® Recombinant Proteins includes cell proliferation, cytotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, and protease assays. Recombinant Human Interleukin-28A (IL28A) specific activity is determined by the dose dependent protection of the cytopathic effect on A549 cells.

Proven compatibility
Gibco® proteins are bioassayed with Gibco® media. Since 1962, Gibco® has been a consistent provider of high quality media, reagents, and sera for reliable cell culture.

Related links
See our selection of recombinant Gibco® growth factors.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Drug Development, Immunology, Virology
Activity: 5 ng/ml
Bioassay: Protection of the cytopathic effect on encephalomyocarditis virus challenged A549 cells
Endotoxin Level: < 1 EU/ml
Expression: Mammalian
Form: Lyophilized
Gene Aliases: IL-28A
Mass: 10 µg
Molecular Weight: 24 kDa
Product Size: 10 µg
Protein Family: Cytokines & Receptors
Protein Form: Ligand, Recombinant
Protein Subtype: Interleukins
Purity: 95 %
Purity or Quality Grade: Carrier-Free
Species: Human
Validated Application: Cell Culture
Shipping Condition: Dry Ice

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products