MIP-1-beta Recombinant Human Protein

Catalog number: PHC1031L

Gibco™  Related applications: Mammalian Cell Culture | Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant MIP-1-beta is a bioactive protein intended for use in cell culture applications. MIP-1&beta (Macrophage inflammatory protein-1&alpha) is involved in a number of biological processes including chemotaxis and is involved in inducing the synthesis of pro-inflammatory cytokines including TNF, IL-6, and IL-1.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Immunology, Inflammation, Neurobiology, Virology
Endotoxin Level: < 0.1 ng/µg
Expression: E. coli
Form: Liquid
Gene Aliases: MIP-1β
Gene ID: 6351
Mass: 100 µg
Molecular Weight: 8 kDa
Product Line: Gibco®
Product Size: 100 µg
Protein Family: Chemokines & Receptors
Protein Form: Ligand, Recombinant
Protein Subtype: MIP (Macrophage Inflammatory Protein) Chemokines
Purification: Sequential Chromatography
Purity: 95 %
Purity or Quality Grade: Carrier-Free
Species: Human
Validated Application: Cell Culture
Shipping Condition: Dry Ice

Contents & storage

Store in ultra-cold freezer (-68 to -85°C).

Documents

Manuals & protocols

Recommended products