CXCL1 (GROα) Recombinant Human Protein

Catalog number:  PHC1063

Gibco™  Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant Human Chemokine (C-X-C motif) ligand 1 (Also known as CXCL1, GROα, melanoma growth stimulating activity, MSGA, Neutrophil-activating protein 3, NAP-3) is a bioactive protein intended for use in cell culture applications. CXCL1 is a chemokine that induces the infiltration of neutrophils.

High purity—no interference from other proteins or contaminants
High biological activity—more results with less protein
Proven compatibility—Gibco® proteins bioassayed with Gibco® media

High Purity Recombinant Bioactive Protein
Recombinant Human Chemokine (C-X-C motif) ligand 1 is one of our more than 250 available Gibco® recombinant proteins. To help ensure Gibco® growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein.

High Biological Activity
In-house activity testing of Gibco® Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, antiviral, and protease assays. Recombinant Human Chemokine (C-X-C motif) ligand 1 (CXCL1, GROα) specific activity is determined by measuring the dose dependent mobilization of intracellular calcium (calcium flux) with human neutrophils.

Proven Compatibility
Gibco® proteins are bioassayed with Gibco® media. Since 1962, Gibco® has been the consistent provider of high quality media, reagents, and sera for reliable cell culture.

See our selection of recombinant Gibco® growth factors.

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Angiogenesis, Cell Cycle & Proliferation, Inflammation, Oncology, Stem Cell Research
Endotoxin Level: < 0.1 ng/µg
Expression: E. coli
Form: Lyophilized
Gene Aliases: CXCL1 (GROα)
Gene ID: 2919
Mass: 1 mg
Molecular Weight: 8 kDa
Product Line: Gibco®
Product Size: 1 mg
Protein Family: Chemokines & Receptors
Protein Form: Ligand, Recombinant
Protein Subtype: GRO⁄KC⁄CINC Chemokines
Purification: Sequential Chromatography
Purity: 95 %
Purity or Quality Grade: Carrier-Free
Species: Human
Validated Application: Cell Culture
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products