Cripto Recombinant Human Protein

Catalog number: PHC8013

Gibco™  Related applications: Cell Viability, Proliferation & Function | Stem Cell Culture | Stem Cell Differentiation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant human Cripto is a bioactive protein intended for use in cell culture applications. Cripto is a member of the EGF-related family of growth factors.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Differentiation, Stem Cell Research
Accession Number: AK163969, BAE21807
Endotoxin Level: Very Low
Expression: Mammalian
Form: Lyophilized
Gene Aliases: Cripto
Gene ID: AK163969, BAE21807
Gene Symbol: TDGF1
Mass: 1 mg
Molecular Weight: 17.8 kDa
Product Line: Gibco®
Product Size: 1 mg
Protein Family: Growth Factors & Receptors
Protein Form: Full Length, Ligand, Recombinant
Protein Length or Portion (a.a.): 139 aa
Protein Subtype: Other Proteins
Protein Tag or Fusion: Untagged
Purification: Sequential Chromatography
Purity: 95 %
Purity or Quality Grade: Carrier-Free
Species: Human
Validated Application: Cell Culture
Shipping Condition: Wet Ice

Contents & storage

This product is supplied lyophilized, carrier-free.
Store lyophilized human Cripto-1 at 2 to 8°C, preferably desiccated. Upon reconstitution, apportion into working aliquots and store at = –20°C. Avoid repeated freeze⁄thaw cycles.

Documents

Manuals & protocols

Recommended products