Cripto Recombinant Human Protein

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant human Cripto is a bioactive protein intended for use in cell culture applications. Cripto is a member of the EGF-related family of growth factors.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Lyophilized
Mass: 10 µg
Purity: 95 %
Gene ID: 4507424, 57932
Species: Human
Expression: Mammalian
Gene Aliases: Cripto
Product Size: 10 µg
Protein Form: Ligand, Recombinant
Purification: Sequential Chromatography
Protein Family: Growth Factors & Receptors
Endotoxin Level: < 0.1 ng/µg
Protein Subtype: EGF (Epidermal Growth Factor)
Molecular Weight: 17.8 kDa
Research Category: Differentiation, Stem Cell Research
Validated Application: Cell Culture
Purity or Quality Grade: Carrier-Free
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products