BMPR1A Recombinant Human Protein

Catalog number: PHC9241

Gibco™  Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Bone Morphogenetic Protein Receptor 1A (BMPR1A) Fc Chimera is a bioactive protein intended for use in cell culture applications. BMPR1A is one of the bone morphogenetic protein (BMP) receptors which are a family of transmembrane serine/threonine kinases that include the type I receptors BMPR1A and BMPR1B and the type II receptor BMPR2. These receptors are also closely related to the activin receptors, ACVR1 and ACVR2. The ligands of these receptors are members of the TGF-beta superfamily. TGF-betas and activins transduce their signals through the formation of heteromeric complexes with 2 different types of serine (threonine) kinase receptors: type I receptors of about 50-55 kD and type II receptors of about 70-80 kD. Type II receptors bind ligands in the absence of type I receptors, but they require their respective type I receptors for signaling, whereas type I receptors require their respective type II receptors for ligand binding.

High purity—no interference from other proteins or contaminants
High biological activity—more results with less protein
Proven compatibility—Gibco® proteins bioassayed with Gibco® media

High Purity Recombinant Bioactive Protein
Bone Morphogenetic Protein Receptor 1A Fc Chimera is one of our more than 250 available Gibco® recombinant proteins. To help ensure Gibco® growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein.

High Biological Activity
In-house activity testing of Gibco® Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, antiviral, and protease assays. Bone Morphogenetic Protein Receptor 1A (BMPR1A) specific activity is determined by its ability to inhibit human BMP4induction of alkaline phosphatase in the mouse chondrogenic cell line ATDC5.

Proven Compatibility
Gibco® proteins are bioassayed with Gibco® media. Since 1962, Gibco® has been the consistent provider of high quality media, reagents, and sera for reliable cell culture.

See our selection of recombinant Gibco® growth factors.

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Stem Cell Research
Accession Number: NM_004329
Biological Activity: ED50 range = 0.1 to 0.3 µg/ml
Endotoxin Level: < 0.1 ng/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: BMPR1A
Gene ID: BMPR1a
Indicator Cell: mouse chondrogenic cell line ATDC5
Mass: 100 µg
Molecular Weight: 43.7 kDa
Product Line: Gibco®
Product Size: 100 µg
Protein Family: Growth Factors & Receptors
Protein Form: Fc Chimera, Ligand, Recombinant
Protein Subtype: BMPs (Bone Morphogenetic Proteins) & Related Factors
Protein Tag or Fusion: Untagged
Purification: Sequential Chromatography
Purity: 95 %
Purity or Quality Grade: Carrier-Free
Reactivity: Human
Species: Human
Validated Application: Cell Culture
Shipping Condition: Wet Ice

Contents & storage

Store lyophilized human BMPR1a at 2 to 8°C, preferably desiccated. Upon reconstitution, apportion into working aliquots and store at ≤ –20°C. Avoid repeated freeze⁄thaw cycles.

Documents

Manuals & protocols

Recommended products