IL33 Recombinant Human Protein

Catalog number: PHC9254

Gibco™  Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant IL33 is a bioactive protein intended for use in cell culture applications. IL33 is a proinflammatory cytokine with structural homology similar with IL-1 family cytokines. IL33 exerts its biological effects by binding to the receptor ST2 and forming a complex with IL-1 Receptor Accessory Protein. Intracellular proteins within in the NF-κB and MAP kinase signaling pathways are activated leading to the production of the type 2 cytokines IL-5 from polarized TH2 cells.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Cell Cycle & Proliferation, Signal Transduction
Accession Number: NM_033439
Bioassay: Proliferation of Mouse D10S Cells
Biological Activity: ED50 range = 0.1-1.0 ng⁄mL
Endotoxin Level: < 0.1 ng/µg
Expression: E. coli
Form: Lyophilized
Gene Aliases: IL-33
Gene ID: 90865
Gene Symbol: IL33
Mass: 10 µg
Molecular Weight: 18 kDa
Product Line: Gibco®
Product Size: 10 µg
Protein Family: Cytokines & Receptors
Protein Form: Ligand, Recombinant
Protein Length or Portion (a.a.): 159 aa
Protein Subtype: Interleukins
Purification: Sequential Chromatography
Purity: 95 %
Purity or Quality Grade: Carrier-Free
Reactivity: Human
Species: Human
Specific Activity: 1.0 x 107 - 1.0 x 106 units⁄mg
Validated Application: Cell Culture
Shipping Condition: Wet Ice

Contents & storage

Lyophilized Human IL-33 should be stored at 2-8°C, preferably desiccated. Store reconstituted Human IL-33 at ≤ –20°C (not in a frost-free freezer). Keep freeze-thaw cycles to a minimum.
Please look at all pack sizes when trying to fill an order.

Documents

Manuals & protocols

Recommended products