IL31 Recombinant Human Protein

Catalog number: PHC9264

Gibco™  Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant IL31 is a bioactive protein intended for use in cell culture applications. IL31 is primarily associated with activated T cells and expressed by Th2 rather than Th1 cells. IL31 signals via IL31RA and the oncostatin M receptor. IL31 signaling has been shown to involve the Jak/STAT pathway, the PI3 kinase/AKT cascade, and MAP kinase pathway.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Immunology, Inflammation
Endotoxin Level: < 0.1 ng/µg
Expression: E. coli
Form: Lyophilized
Gene Aliases: IL-31
Mass: 10 µg
Molecular Weight: 16 kDa
Product Line: Gibco®
Product Size: 10 µg
Protein Family: Cytokines & Receptors
Protein Form: Ligand, Recombinant
Protein Subtype: Interleukins
Purification: Sequential Chromatography
Purity: >95% by SDS-PAGE
Purity or Quality Grade: Carrier-Free
Reactivity: Human
Species: Human
Validated Application: Cell Culture
Shipping Condition: Wet Ice

Contents & storage

Lyophilized Human IL-31 should be stored at 2-8°C, preferably desiccated. Store reconstituted Human IL-31 at ≤ –20°C (not in a frost-free freezer). Keep freeze-thaw cycles to a minimum.

Documents

Manuals & protocols

Recommended products