IL29 Recombinant Human Protein

Catalog number:  PHC9274

Gibco™  Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant IL29 is a bioactive protein intended for use in cell culture applications. IL29 is functionally similar to type I interferons, including antiviral activity and up-regulation of MHC class I antigen expression. IL29 signal through the same heterodimeric receptor complex that is composed of the IL-10 receptor b (IL-10 Rb) and a novel IL-28 receptor a (IL-28 Ra, also known as IFN-γ R1). Binding to the receptor complex induces Jak kinase activation, as well as STAT1 and STAT2 tyrosine phosphorylation.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Immunology, Virology
Endotoxin Level: < 0.1 ng/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: IL-29
Mass: 10 µg
Molecular Weight: 35 kDa
Product Line: Gibco®
Product Size: 10 µg
Protein Family: Cytokines & Receptors
Protein Form: Ligand, Recombinant
Protein Subtype: Interleukins
Purification: Sequential Chromatography
Purity: >95% by SDS-PAGE
Purity or Quality Grade: Carrier-Free
Reactivity: Human
Species: Human
Validated Application: Cell Culture
Shipping Condition: Wet Ice

Contents & storage

Lyophilized Human IL-29 should be stored at 2-8°C, preferably desiccated. Store reconstituted Human IL-29 at ≤ –20°C (not in a frost-free freezer). Keep freeze-thaw cycles to a minimum.

Documents

Manuals & protocols

Recommended products