Activin R1b Recombinant Human Protein

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant Activin R1b is a bioactive protein intended for use in cell culture applications. Recombinant Activin Receptor 1B (ActivinR1B) is intended for use in cell culture applications. ActivinR1B is involved in a number of biological processes including growth and differentiation, cell proliferation, apoptosis, and embryonic cell fate.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Differentiation, Stem Cell Research
Accession Number: NM_004302
Biological Activity: Linear range of 0.8 to 100 ng⁄mL
Endotoxin Level: < 0.1 ng/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: Activin R1b
Gene ID: 91
Gene Symbol: ACVR1B
Mass: 1 mg
Molecular Weight: 38.2 kDa
Product Line: Gibco®
Product Size: 1 mg
Protein Family: Growth Factors & Receptors
Protein Form: Ligand, Recombinant
Protein Length or Portion (a.a.): 343 aa
Protein Subtype: Activins
Purification: Sequential Chromatography
Purity: 95 %
Purity or Quality Grade: Carrier-Free
Reactivity: Human
Species: Human
Validated Application: Cell Culture
Shipping Condition: Wet Ice

Contents & storage

Lyophilized human ACVR1B should be stored at 2-8°C, preferably desiccated. Store reconstituted human ACVR1B at ≤ –20°C (not in a frost-free freezer). Keep freeze-thaw cycles to a minimum.
Please look at all pack sizes when trying to fill an order.

Documents

Manuals & protocols

Recommended products