Kallikrein 7 Recombinant Human Protein

Catalog number: PHC9295

Gibco™  Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant Kallikrein 7 is a bioactive protein intended for use in cell culture applications. KLK7 is one of the Kallikreins, which are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Lyophilized
Mass: 25 µg
Purity: 95 %
Gene ID: KLK7
Species: Human
Expression: Mammalian
Reactivity: Human
Gene Aliases: Kallikrein 7
Product Size: 25 µg
Protein Form: Recombinant, Ligand
Purification: Sequential Chromatography
Protein Family: Growth Factors & Receptors
Endotoxin Level: < 0.1 ng/µg
Protein Subtype: TGFs (Transforming Growth Factors)
Accession Number: NM_005046
Molecular Weight: 30 kDa
Research Category: Oncology, Dermatology, Inflammation
Specific Activity: 300pmoles⁄min⁄µg
Protein Tag or Fusion: Untagged
Validated Application: Cell Culture
Purity or Quality Grade: Carrier-Free
Shipping Condition: Wet Ice

Contents & storage

Lyophilized Human Kallikrein 7 should be stored at 2-8°C, preferably desiccated. Store reconstituted Human Kallikrein 7 at ≤ –20°C (not in a frost-free freezer). Keep freeze-thaw cycles to a minimum.

Documents

Manuals & protocols

Recommended products