Kallikrein 11 Recombinant Human Protein

Catalog number: PHC9315

Gibco™  Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant Kallikrein 11 is a bioactive protein intended for use in cell culture applications. KLK11 is one of the Kallikreins, which are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Gene Regulation, Oncology
Accession Number: NM133775
Endotoxin Level: < 0.1 ng/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: Kallikrein 11
Gene ID: KLK11
Mass: 25 µg
Molecular Weight: 25.6 kDa
Product Line: Gibco®
Product Size: 25 µg
Protein Family: Cytokines & Receptors
Protein Form: Ligand, Recombinant
Protein Length or Portion (a.a.): 250 aa
Protein Subtype: Other Proteins
Purification: PAGE
Purity: 95 %
Purity or Quality Grade: Carrier-Free
Reactivity: Human
Species: Human
Specific Activity: 2200 pmoles⁄min⁄µg
Validated Application: Cell Culture
Shipping Condition: Wet Ice

Contents & storage

Lyophilized Human Kallikrein 11 should be stored at 2-8°C, preferably desiccated. Store reconstituted Human Kallikrein 11 at ≤ –20°C (not in a frost-free freezer). Keep freeze-thaw cycles to a minimum.

Documents

Manuals & protocols

Recommended products