Kallikrein 4 Recombinant Human Protein

Catalog number: PHC9345

Gibco™  Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant Kallikrein 4 is a bioactive protein intended for use in cell culture applications. KLK4 is one of the Kallikreins, which are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Neurobiology
Endotoxin Level: < 0.1 ng/µg
Expression: Mammalian
Form: Lyophilized
Gene Aliases: Kallikrein 4
Mass: 25 µg
Molecular Weight: 25 kDa
Product Line: Gibco®
Product Size: 25 µg
Protein Family: Cytokines & Receptors
Protein Form: Ligand, Recombinant
Protein Subtype: Other Proteins
Purification: Sequential Chromatography
Purity: >95% by SDS-PAGE
Purity or Quality Grade: Carrier-Free
Reactivity: Human
Species: Human
Specific Activity: > 3000 pmoles⁄min⁄µg
Validated Application: Cell Culture
Shipping Condition: Wet Ice

Contents & storage

Lyophilized Human Kallikrein 4 should be stored at 2-8°C, preferably desiccated. Store reconstituted Human Kallikrein 4 at ≤ -20°C (not in a frost-free freezer). Keep freeze-thaw cycles to a minimum.

Documents

Manuals & protocols

Recommended products