Frizzled 4 Recombinant Human Protein

Catalog number: PHC9431

Gibco™  Related applications: Cell Viability, Proliferation & Function | Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant Frizzled 4 is a bioactive protein intended for use in cell culture applications. FZD4 is a member of the Frizzled family, which are 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. Wnt signaling is involved in several developmental processes.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Endotoxin Level: < 0.1 ng/µg
Expression: Mammalian
Form: Lyophilized
Molecular Weight: 53 kDa
Product Line: Gibco®
Product Size: 100 µg
Protein Form: Recombinant
Protein Subtype: Wnt Proteins & Receptors
Purification: Sequential Chromatography
Purity: >95% by SDS-PAGE
Reactivity: Human
Research Category: Development, Signal Transduction
Species: Human
Validated Application: Cell Culture
Shipping Condition: Wet Ice

Contents & storage

Lyophilized Human Frizzled 4 should be stored at 2-8°C, preferably desiccated. Store reconstituted Human Frizzled 4 at ≤ -20°C (not in a frost-free freezer). Keep freeze-thaw cycles to a minimum.

Documents

Manuals & protocols