Gibco™

Frizzled 7 Recombinant Human Protein

Catalog number:  PHC9474

 Related applications: Cell Viability, Proliferation & Function | Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant Human Frizzled-7 (FZD7) is a bioactive recombinant protein intended for use in cell culture applications. FZD7 is a member of the Frizzled family, which are 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. Wnt signaling is involved in several developmental processes.

The protein sequence of Frizzled-7 corresponds to the extracellular region of mature Human Frizzled 7, aa 33-185, fused to the Fc region of Human IgG1.

The equilibrium binding constant, KD, of Human Frizzled 7 was measured at less than 10nM using mouse Wnt3a as the ligand. Measurements were made using the ForteBio Octet optical biosensor instrument.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Biological Activity: KD = <10 nM
Endotoxin Level: < 0.1 ng/µg
Expression: Mammalian
Form: Lyophilized
Mass: 10 µg
Molecular Weight: 98 kDa
Product Line: Gibco®
Product Size: 10 µg
Protein Form: Recombinant
Protein Length or Portion (a.a.): aa 33-185
Protein Subtype: Wnt Proteins & Receptors
Purification: Sequential Chromatography
Purity: >95 %
Purity or Quality Grade: Carrier-Free
Research Category: Development, Signal Transduction
Species: Human
Validated Application: Cell Culture
Shipping Condition: Wet Ice

Contents & storage

Store this lyophilized preparation at 2 to 8°C. Upon reconstitution, apportion into working aliquots and store at ≤ –20°C. Avoid repeated freeze⁄thaw cycles.

Documents

Manuals & protocols