IL17F Recombinant Human Protein

Catalog number: PHC9554

Gibco™  Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant Human IL17F is a bioactive protein intended for use in cell culture applications. IL17F is a member of structurally related cytokines. The IL-17 family of cytokines plays a role in mediating proinflamatory responses. Recombinant Human IL17F is one of the many GIBCO® recombinant bioactive proteins available from Life Technologies. This is the 10 microgram size.

High purity—minimal interference from other proteins or contaminants
High biological activity—more results with less protein
Proven compatibility—GIBCO® proteins bioassayed with GIBCO® media

High purity recombinant bioactive protein
Recombinant human IL17F is one of more than 250 GIBCO® recombinant proteins available from Life Technologies. To help ensure GIBCO® growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to help ensure a biologically active protein.

High biological activity
In house activity testing includes a variety of cell proliferation, cytotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, and protease assays. Recombinant IL17F specific activity is determined by the dose-dependent induction of IL-6 secretion from NDHF Adult fibroblasts.

Proven Compatibility with GIBCO® Media
GIBCO® proteins are bioassayed with GIBCO® media. Since 1962 GIBCO® has been a consistent provider of high quality media, reagents, and sera for reliable cell culture.

We also offer a selection of growth factors for use in cell therapy applications through our GIBCO® Cell Therapy Systems (CTS™) product line.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

Related links
See our selection of recombinant GIBCO® growth factors.
Learn more about the GIBCO® Cell Therapy Systems (CTS™) product line.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Angiogenesis, Immunology, Inflammation, Neurobiology, Signal Transduction
Activity: 100 ng/ml
Bioassay: Induction of IL-6 secretion from NDHF cells
Endotoxin Level: < 1 EU/ml
Expression: Mammalian
Gene Aliases: IL-17F
Molecular Weight: 38 kDa
Product Line: Gibco®
Product Size: 10 µg
Protein Family: Cytokines & Receptors
Protein Form: Ligand, Recombinant
Protein Length or Portion (a.a.): 133 aa
Protein Subtype: Interleukins
Purification: Gel-Purified
Purity: 90 %
Purity or Quality Grade: Carrier-Free
Species: Human
Validated Application: Cell Culture
Shipping Condition: Dry Ice

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products