IGFBP4 Recombinant Human Protein

Catalog number: PHC9611

Gibco™  Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant Human Insulin-like Growth Factor Binding Protein 4 (IGFBP4) is a bioactive protein intended for use in cell culture applications. IGFBP4 binds IGF1 and II, prolonging these growth factors' half life and altering the way these growth factors interact with cells surface receptors. This is the 100 microgram size.

• High purity—Minimizes interference from other proteins or contaminants
• High biological activity—More results with less protein
• Proven compatibility—Gibco® proteins bioassayed with Gibco® media

High Purity Recombinant Bioactive Protein
Recombinant Human Insulin-like Growth Factor Binding Protein 4 (IGFBP4) is one of our more than 250 available Gibco® recombinant proteins. To help ensure Gibco® growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to help ensure a biologically active protein.

High Biological Activity
In-house activity testing of Gibco® Recombinant Proteins includes cell proliferation, cytotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, and protease assays. Recombinant Human Insulin-like Growth Factor Binding Protein 4 (IGFBP4) specific activity is determined the dose dependent inhibition of IGF-II induced proliferation of MCF-7 cells.

Proven Compatibility
Gibco® proteins are bioassayed with Gibco® media. Since 1962, Gibco® has been a consistent provider of high quality media, reagents, and sera for reliable cell culture.

Related Links
See our selection of recombinant Gibco® growth factors.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Apoptosis, Oncology
Activity: 0.09 µg/ml
Bioassay: inhibition of MCF-7 cell proliferation in the presence of 14 ng/mL IGF2
Endotoxin Level: < 1 EU/ml
Expression: Mammalian
Form: Lyophilized
Gene Aliases: IGFBP-4
Mass: 100 µg
Molecular Weight: 26 kDa
Product Line: Gibco®
Product Size: 100 µg
Protein Family: Growth Factors & Receptors
Protein Form: Ligand, Recombinant
Protein Subtype: IGFs (Insulin-Like Growth Factors)
Purity: 95 %
Purity or Quality Grade: Carrier-Free
Species: Human
Validated Application: Cell Culture
Shipping Condition: Wet Ice

Contents & storage

Store at 2 to 8°C

Documents

Manuals & protocols

Recommended products