Vitronectin Human Protein, Natural

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Natural Human Vitronectin is a bioactive protein intended for use in cell culture applications. Natural human vitronectin is intended for use in cell culture applications. Vitronectin is involved in cell adhesion, migration, proliferation, and differentiation of some cell types.

High purity—no interference from other proteins or contaminants
High biological activity—more results with less protein
Proven compatibility—Gibco® proteins bioassayed with Gibco® media

High Purity Recombinant Bioactive Protein
Natural Human Vitronectin is one of our more than 250 available Gibco® recombinant proteins. To help ensure Gibco® growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein.

High Biological Activity
In-house activity testing of Gibco® Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, antiviral, and protease assays. Natural Human Vitronectin specific activity is determined by measuring the dose dependent attachment of TIG-3 cells to Vitronectin coated plates.

Proven Compatibility
Gibco® proteins are bioassayed with Gibco® media. Since 1962, Gibco® has been the consistent provider of high quality media, reagents, and sera for reliable cell culture.

See our selection of recombinant Gibco® growth factors.

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Extracellular Matrix & Cell Adhesion, Stem Cell Research
Species: Human
Protein Subtype: Vitronectin
Validated Application: Cell Culture
Gene Aliases: Vitronectin
Product Line: Gibco®
Product Size: 100 µg
Shipping Condition: Dry Ice

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products