Gibco™

Fibronectin Human Protein, Native

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Natural Human Fibronectin is a bioactive protein intended for use in cell culture applications. Natural human fibronectin is intended for use in cell culture applications. Fibronectin is involved in a number of cellular processes including cell adhesion, migration, morphology, and regulation of cell growth and differentiation.

High purity—no interference from other proteins or contaminants
High biological activity—more results with less protein
Proven compatibility—Gibco® proteins bioassayed with Gibco® media

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Extracellular Matrix & Cell Adhesion, Stem Cell Research
Species: Human
Protein Subtype: Fibronectin
Validated Application: Cell Culture
Gene Aliases: Fibronectin Natural Human
Product Line: Gibco®
Product Size: 1 mg
Shipping Condition: Dry Ice

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols