IL6 Recombinant Mouse Protein

Catalog number: PMC0066

Gibco™  Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant Mouse Interleukin 6 (IL6) is a bioactive protein intended for use in cell culture applications. IL6 is a pleiotropic cytokine involved in several cellular processes including inflammation, hematopoeisis, and immune response.

High purity—no interference from other proteins or contaminants
High biological activity—more results with less protein
Proven compatibility—Gibco® proteins bioassayed with Gibco® media

High Purity Recombinant Bioactive Protein
Recombinant Mouse Interleukin 6 is one of our more than 250 available Gibco® recombinant proteins. To help ensure Gibco® growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein.

High Biological Activity
In-house activity testing of Gibco® Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, antiviral, and protease assays. Recombinant Mouse Interleukin 6 (IL6) specific activity is determined by the dose dependent proliferation of mouse B9 cells.

Proven Compatibility
Gibco® proteins are bioassayed with Gibco® media. Since 1962, Gibco® has been the consistent provider of high quality media, reagents, and sera for reliable cell culture.

See our selection of recombinant Gibco® growth factors.

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Lyophilized
Mass: 25 µg
Purity: 95 %
Gene ID: 16193
Species: Mouse
Expression: E. coli
Gene Aliases: IL-6
Product Size: 25 µg
Protein Form: Recombinant, Ligand
Purification: Sequential Chromatography
Protein Family: Cytokines & Receptors
Endotoxin Level: < 0.1 ng/µg
Protein Subtype: Interleukins
Molecular Weight: ~20 kDa
Research Category: Immunology, Stem Cell Research, Inflammation, Bone Research
Validated Application: Cell Culture
Purity or Quality Grade: Carrier-Free
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products