SCF (C-Kit Ligand) Recombinant Mouse Protein

Catalog number: PMC2113L

Gibco™  Related applications: Cell Viability, Proliferation & Function | Mammalian Cell Culture | Stem Cell Culture | Stem Cell Differentiation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant Mouse Stem Cell Factor (SCF, C-Kit Ligand) is a bioactive protein intended for use in cell culture applications. SCF is a pleiotropic factor that acts in utero in germ cell and neural cell development, and hematopoiesis, all believed to reflect a role in cell migration. In adults, it functions pleiotropically, while mostly noted for its continued requirement in hematopoiesis.

High purity—no interference from other proteins or contaminants
High biological activity—more results with less protein
Proven compatibility—Gibco® proteins bioassayed with Gibco® media

High Purity Recombinant Bioactive Protein
Recombinant Mouse Stem Cell Factor is one of our more than 250 available Gibco® recombinant proteins. To help ensure Gibco® growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein.

High Biological Activity
In-house activity testing of Gibco® Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, antiviral, and protease assays. Recombinant Mouse Stem Cell Factor (SCF, C-Kit Ligand) specific activity is determined by the dose dependent proliferation of human MO-7e cells.

Proven Compatibility
Gibco® proteins are bioassayed with Gibco® media. Since 1962, Gibco® has been the consistent provider of high quality media, reagents, and sera for reliable cell culture.

See our selection of recombinant Gibco® growth factors.

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Differentiation, Oncology, Stem Cell Research
Endotoxin Level: < 0.1 ng/µg
Expression: E. coli
Form: Liquid
Gene Aliases: SCF (c-kit Ligand)
Mass: 1 mg
Molecular Weight: 18.5 kDa
Product Line: Gibco®
Product Size: 1 mg
Protein Family: Cytokines & Receptors
Protein Form: Ligand, Recombinant
Protein Subtype: Other Proteins
Purification: Sequential Chromatography
Purity: 95 %
Purity or Quality Grade: Carrier-Free
Species: Mouse
Validated Application: Cell Culture
Shipping Condition: Dry Ice

Contents & storage

Store in ultra-cold freezer (-68 to -85°C).

Documents

Manuals & protocols

Recommended products