Gibco™

TNF Recombinant Rat Protein

Catalog number:  PRC3014

 Related applications: Cell Viability, Proliferation & Function | Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant TNF is a bioactive protein intended for use in cell culture applications. TNF causes cytolysis and cytostasis of many tumor cell lines. TNF has a wide spectrum of activities, including chemotaxis of neutrophils, alteration of the endothelium, inhibition of anticoagulatory mechanisms, and promotion of angiogenesis.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Immunology, Inflammation, Oncology, Signal Transduction, Stem Cell Research
Endotoxin Level: < 0.1 ng/µg
Expression: E. coli
Form: Lyophilized
Gene Aliases: TNF-α
Gene ID: 24835
Mass: 10 µg
Molecular Weight: 17 kDa
Product Line: Gibco®
Product Size: 10 µg
Protein Family: Cytokines & Receptors
Protein Form: Ligand, Recombinant
Protein Subtype: TNF (Tumor Necrosis Factor)
Purification: Sequential Chromatography
Purity: 95 %
Purity or Quality Grade: Carrier-Free
Species: Rat
Validated Application: Cell Culture
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products