Gibco™

VEGF Recombinant Rat Protein

Catalog number:  PRG0113

 Related applications: Cell Viability, Proliferation & Function | Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant VEGF is a bioactive protein intended for use in cell culture applications. VEGF exerts angiogenic, mitogenic and vascular permeability-enhancing activities specific for endothelial cells. VEGF has also been shown to be chemotactic for monocytes and osteoblasts.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Research Category: Angiogenesis, Inflammation, Oncology, Stem Cell Research
Endotoxin Level: < 0.1 ng/µg
Expression: E. coli
Form: Lyophilized
Gene Aliases: VEGF
Mass: 1 mg
Molecular Weight: 38.4 kDa
Product Line: Gibco®
Product Size: 1 mg
Protein Family: Growth Factors & Receptors
Protein Form: Ligand, Recombinant
Protein Subtype: VEGF (Vascular Endothelial Growth Factor)
Purification: Exchange Chromatography
Purity: 95 %
Purity or Quality Grade: Carrier-Free
Species: Rat
Validated Application: Cell Culture
Shipping Condition: Approved for shipment on Wet or Dry Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products