Thermo Scientific™
Prima PRO Process Mass Spectrometer
 Related applications: Industrial Mass Spectrometry
Maximize product yield and increase profits with the Thermo Scientific™ Prima PRO Process Mass Spectrometer. This process gas analyzer is engineered to meet a number of challenging process applications in the petrochemical, iron and steel, and biotechnology industries. Highly reliable and easy-to-own, the Prima PRO delivers faster, more complete, lab quality online gas composition analysis. It features a rugged, fault-tolerant design that ensures availability that exceeds 99.7% and a simplified maintenance procedure and auto-tune capability to facilitate ongoing operation and enhance productivity.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Features:
  • Significantly enhanced analyzer control, giving increased sensitivity, precision, accuracy and mass range
  • Completely redesigned electronics, incorporating higher specification components
  • Fast online process gas analysis (1 to 20 seconds per point) for accurate tracking of process dynamics
  • Comprehensive measurement with more data provided to advanced process control (APC) models
  • Stable with 30- to 90-day automated calibration interval
  • Reliable, fault-tolerant design for availability of greater than 99.7%
  • Complete service access from the front of the system, reducing the effective footprint
  • Small footprint with no large shelter required
  • Minimal maintenance requirements
  • Enhanced connectivity to DCS systems and off-line data manipulation packages
  • Thermo Scientific™ GasWorks Software
Recommended for:
  • Fermentation processes
  • Cell culture processes
  • Iron and steel processes
  • Ethylene, Ammonia, Hydrogen plants
  • Ethylene Oxide, Methanol, Polyethylene/Polypropylene
  • Catalysis research
  • Solvent drying
access the user manuals for mass spectrometry instruments
To access the user manuals, please contact us directly:
email: Techsupport.epm.europe@thermofisher.com
phone:  +44 1606 548 753
Documents
Manuals & protocols