PowerEase™ 90W Power Supply (230 VAC) (Australia)

Catalog number:  PS0092

 Related applications: Protein Assays and Analysis | Protein Gel Electrophoresis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The PowerEase® 90W Power Supply is designed specifically for mini-gel electrophoresis. The straight-forward, intuitive interface makes the powering of DNA and protein gel runs a simple and easy process.

In addition, the PowerEase® 90W Power Supply features:

• Constant voltage or current settings
• Built-in timer for walk-away gel electrophoresis
• Four sets of output jacks that are compatible with most electrophoresis devices

Easy to Use
With the PowerEase® 90W Power Supply, simply set a constant voltage or current to begin a run. This power supply easily accommodates the running and transferring of 2 mini-gels, such as Bolt® Bis-Tris Plus gels or 2 midi-gels using the XCell4 SureLock™ Midi-Cell.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Novex® Semi-Dry Blotter, XCell II Blot Module, XCell SureLock® Mini, XCell4 SureLock™ Midi, XCell6™ MultiGel
Product Line: PowerEase®
Product Size: 1 each
Type: Power Supply

Contents & storage

The PowerEase® 90W Power Supply includes the power supply and a quick reference card.

Documents

Manuals & protocols