IFN-γ Recombinant Swine Protein

Catalog number: PSC4035

Gibco™  Related applications: Cell Viability, Proliferation & Function | Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Recombinant IFN-γ is a bioactive protein intended for use in cell culture applications. Recombinant swine IFN-γ is intended for use in cell culture applications. IFN-γ is involved in many biological processes such as inducing the secretion of TNF-α and stimulating the release of reactive oxygen species from macrophages. It is also involved with bone growth and inhibits bone resorption. It is antiviral and antiparasitic and inhibits the proliferation of cells.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Lyophilized
Mass: 25 µg
Purity: 95 %
Gene ID: 396991
Species: Swine⁄Porcine
Expression: E. coli
Gene Aliases: IFN-γ
Product Size: 25 µg
Protein Form: Recombinant, Ligand
Purification: Sequential Chromatography
Protein Family: Cytokines & Receptors
Endotoxin Level: < 0.1 ng/µg
Protein Subtype: Interferons
Molecular Weight: 17.1 kDa
Research Category: Immunology, Oncology, Angiogenesis, Virology, Signal Transduction
Validated Application: Cell Culture
Purity or Quality Grade: Carrier-Free
Shipping Condition: Approved for shipment on Wet or Dry Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products