Fluorescein-Crosstide Substrate

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

A general substrate for Ser/Thr kinases, the Crosstide peptide substrate is based upon GSK-3alpha/beta.

For use in combination with the LanthaScreen® Tb-pCrosstide Antibody (PV3564) to evaluate inhibitors in medium to high throughput screening applications.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Amino Acid Sequence: Fluorescein- GRPRTSSFAEG
Detection Method: Fluorescent
Gene ID: 10000, 10298, 207, 208, 25400, 51347, 5566, 56164, 57144, 6197, 6198, 8536
Label or Dye: FITC (Fluorescein)
Molecular Weight: 1524 Da
Product Size: 1 mg
Substrate: Fluorescein-Crosstide
Substrate Properties: Peptide-Based Substrate
Substrate Type: Kinase Substrate
Target Enzyme: Serine⁄Threonine Kinases
Validated Kinases: AKT1, AKT2, AKT3, CAMK2A, CAMK2D, PAK1, PAK4, PAK7, PRKACA, RPS6KB1, RPS6KA3, TAOK3
Shipping Condition: Dry Ice

Contents & storage

Fluorescein-Crosstide substrates in 50 mM HEPES, pH 7.5. Store at -20°C.

Documents

Manuals & protocols