LanthaScreen® Tb-PY20 Antibody Kit

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

A general reagent to monitor tyrosine kinase activity, the pY20 antibody binds to phosphorylated tyrosines within a peptide or physiological protein substrate.

For use in combination with Fluorescein-poly-GT (PV3610) or Fluorescein-poly-GAT (PV3611) to evaluate inhibitors in medium to high throughput screening applications.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format: 384-well plate
Readout: End Point
Technique: TR-FRET
Label or Dye: Tb (Terbium), FITC (Fluorescein)
Product Size: 1 mg
Target Enzyme: Tyrosine Kinases
Validated Kinases: ABL1, ABL2, AXL, BLK, BMX, BTK, CSF1R, CSK, DDR2, EGFR, EPHA1, EPHA2, EPHA3, EPHA4, EPHA5, EPHA7, EPHA8, EPHB1, EPHB2, EPHB3, EPHB4, ERBB2, ERBB4, FER, FGFR1, FGFR2, FGFR3, FGFR4, FGR, FLT1, FLT3, FLT4, FRK, FYN, HCK, IGF1R, INSR, KDR, KIT, LCK, LTK, LYNA, LYNB, MATK, MERTK, MET, MST1R, MUSK, NTRK1, NTRK2, NTRK3, PDGFRA, PDGFRB, PTK2, PTK2B, PTK6, RET, ROS1, SRC, SRCN1, SRMS, SYK, TEC, TEK, TNK2, TTK, TYRO3, YES1, ZAP70
Antibody Molecular Weight: 150000 Da
Excitation⁄Emission (nm): 340⁄490, 520
Shipping Condition: Dry Ice

Contents & storage

Terbium-labeled anti-pY20 antibody in HEPES buffered saline. Store at -20°C.

Documents

Manuals & protocols