LanthaScreen® Tb-pCrosstide Antibody Kit

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

A general assay for serine/threonine kinases, the sequence of the Crosstide substrate is based upon GSK-3alpha/beta. The antibody is directed towards the phosphorylated Crosstide substrate.

For use in combination with the Fluorescein-Crosstide peptide substrate (PV3509) to evaluate inhibitors in medium to high throughput screening applications.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format: 384-well plate
Gene ID: 10000, 10298, 207, 208, 25400, 51347, 56164, 57144, 6197, 6198, 8536
Readout: End Point
Technique: TR-FRET
Reactivity: Human
Label or Dye: Tb (Terbium), FITC (Fluorescein)
Product Size: 1 mg
Target Enzyme: Serine⁄Threonine Kinases
Validated Kinases: AKT1, AKT2, AKT3, CAMK2A, CAMK2D, PAK1, PAK4, PAK7, PRKACA, RPS6KB1, RPS6KA3, TAOK3
Antibody Molecular Weight: 150000 Da
Excitation⁄Emission (nm): 340⁄490, 520
Shipping Condition: Dry Ice

Contents & storage

Terbium-labeled anti-Crosstide antibody in HEPES buffered saline. Store at -20°C.

Documents

Manuals & protocols