LanthaScreen® Tb-labeled Anti-Mouse Ab

Catalog number: PV3766

 Related applications: Secondary Detection | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

A general reagent for use in assay development, this terbium labeled antibody will bind mouse antibodies. This product is ideally suited for use during assay development when evaluating multiple primary mouse antibodies or when you are unable to directly terbium label your primary antibody.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reactivity: Mouse
Label or Dye: Tb (Terbium)
Product Size: 25 µg
Shipping Condition: Dry Ice
Excitation⁄Emission (nm): 340⁄490, 520

Contents & storage

Terbium-labeled anti-Mouse antibody in HEPES buffered saline. Store at -20°C.

Documents

Manuals & protocols