For order quantities > 1, multiple lots may be used to fulfill the order. If you require a single lot, please email discoverysciences@lifetech.com to ensure your needs are accommodated

88 more kinases now available to order! See the complete list.

AXL Recombinant Human Protein

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Axl is a chronic myelogenous leukemia-associated oncogene and also associated with colon cancer and melanoma. It transduces signals from the extracellular matrix into the cytoplasm by binding growth factors like vitamin K-dependent protein growth-arrest-specific gene 6, and is involved in the stimulation of cell proliferation.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Gene ID: 558, 7124
Species: Human
Target Gene: AXL receptor tyrosine kinase
Gene Aliases: AXL, UFO
Kinase Group: Tyrosine Kinases
Product Size: 10 µg
Protein Family: Kinases & Inhibitors
Protein Subtype: Receptor Tyrosine Kinases
Accession Number: NP_068713
Molecular Weight: 52.2 kDa
Research Category: Signal Transduction, Oncology
System Compatibility: LanthaScreen® Substrate, LanthaScreen® Antibody, Z'-LYTE™
Protein Tag or Fusion: His Tag
Validated Application: Kinase Assay
Protein Length or Portion (a.a.): aa 473-894
Expression: Baculovirus
Kinase Family: AXL Kinase Family
Protein Form: Recombinant, Cytoplasmic
Shipping Condition: Dry Ice

Contents & storage

Storage Buffer: 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.

Documents

Manuals & protocols

Recommended products