FL - eA2 Peptide, 100µM

Catalog number: PV4610

 Related applications: Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

LanthaScreen® fluorescent nuclear receptor coregulator peptides contain known interaction motifs and are labeled with fluorescein. These peptides are matched and validated to complement the LanthaScreen® TR-FRET Nuclear Receptor Coregulator Assays. Assays developed using these reagents enable primary or secondary screening of nuclear receptor agonists and/or antagonists.

How they work
Ligand binding to nuclear receptors causes conformational changes in the receptor, resulting in a cascade of events, including dissociation of repressor proteins, association of coactivator proteins, and assembly of pol II and other transcriptional factors for activation of target genes. TR-FRET based assays can be developed using the LanthaScreen® panel of fluorescein-labeled coregulator peptides to investigate conformational changes of nuclear receptors upon ligand binding, either by determining the affinity of ligand-bound receptor for different coregulator peptides, or by identifying additional agonists or antagonists via displacement or recruitment of a specific coregulator peptide. In the LanthaScreen® TR-FRET RXR beta Coactivator Assay, a terbium (Tb)-labeled anti-GST antibody is used to indirectly label GSTtagged RXR beta protein. An agonist (9-cis Retinoic Acid) added to the receptor which, upon ligand binding, causes a conformational change resulting in recruitment of the Fl-D22 coactivator peptide (Figure 1). The binding of the fluorescent coregulator peptide to RXR beta causes an increase in the TR-FRET emission ratio (Figure 2).

Contents and Storage:

LanthaScreen® fluorescent nuclear receptor coregulator peptides are supplied as 100 µM solutions in peptide-appropriate buffers. Store peptides at -20°C.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Size: 100 µL
Shipping Condition: Dry Ice

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols