LanthaScreen® Tb-anti-ubiquitin-FK2 Antibody Kit

Catalog number: PV4751

 Related applications: Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Tb-anti-ubiquitin-FK2 antibody preferentially binds to mono- or poly-ubiquitinated proteins over free ubiquitin. This antibody can be used in combination with purified GFP-substrates, fluorescein-ubiquitin, or LanthaScreen® cell lines to develop robust high-throughput biochemical or cell based assays for ubiquitination or deubiquitination.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Antibody Molecular Weight: 150000 Da
Excitation⁄Emission (nm): 340⁄490, 520
For Use With (Equipment): Microplate Reader
Form: Liquid
Format: 384-well plate
Label or Dye: Tb (Terbium)
Product Line: LanthaScreen®
Product Size: 1 mg
Reactivity: Human
Shipping Condition: Dry Ice

Contents & storage

Terbium-labeled anti-ubiquitin-FK2 antibody in HEPES buffered saline. Store at -20°C.

Documents

Manuals & protocols