This kit contains a goat Tb-anti-GST antibody—an equivalent kit containing a rabbit-sourced antibody is also available.

LanthaScreen™ TR-FRET PPAR gamma Competitive Binding Assay Kit, goat

Catalog number:  PV4894

 Related applications: Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This kit contains Goat Tb-Anti-GST antibody; other kit components are the same as kit A15145:

The LanthaScreen® TR-FRET PPAR gamma Competitive Binding Assay provides a sensitive and robust method for high-throughput screening (HTS) of ligands for peroxisome proliferator-activated receptor-gamma (PPAR gamma). The kit uses a terbium-labeled anti-GST antibody, a fluorescent small-molecule pan-PPAR ligand (Fluormone™ Pan-PPAR Green), and human PPAR gamma ligand-binding domain (LBD) that is tagged with glutathione S-transferase (GST) in a homogeneous mix-and-read assay format.

To assay:
When running the LanthaScreen® TR-FRET PPAR gamma Competitive Binding Assay, Fluormone™ Pan-PPAR Green is added to ligand test compounds followed by addition of a mixture of the PPAR gamma-LBD and terbium anti-GST antibody. When the Fluormone™ Pan-PPAR Green is bound to PPAR gamma, energy transfer from the terbium-labeled antibody to the tracer occurs, and a high TR-FRET ratio is observed. Competitive ligand binding to PPAR gamma is detected by a test compound’s ability to displace the tracer, which results in a loss of FRET between the antibody and the tracer. After an incubation period at room temperature, the 520 nm/495 nm TR-FRET ratio is calculated and can be used to determine the IC50 from a dose response curve of the compound (Figure 1). This type of binding assay is analogous to radioligand-based assays, except that it eliminates handling of radioactivity and enables a homogeneous, "addition-only" format.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: Fluorescent
For Use With (Equipment): Microplate Reader
Format: 384-well plate
Label or Dye: Tb (Terbium)
Product Line: LanthaScreen®
Product Size: 400 x 40 µL
Readout: End Point
Technique: TR-FRET
Validated Application: Competitive Binding Assay
Shipping Condition: Dry Ice

Contents & storage

The LanthaScreen® TR-FRET PPAR gamma Competitive Binding Assay Kit contains PPAR gamma-LBD (GST) protein, Fluormone™ Pan-PPAR Green, terbium-labeled anti-GST antibody, and buffers. Store components as indicated in the assay protocol (-80°C, -20°C, or +4°C).

Documents

Manuals & protocols

Recommended products