Kinase Buffer R (5X)

Catalog number: PV5101

 Related applications: Protein Assays and Analysis | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Kinase buffer that has been specially optimized for use with the Class I & II PI3 kinases.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Liquid
Product Size: 4 mL
Shipping Condition: Approved for shipment at Room Temperature or on Wet or Dry Ice

Contents & storage

5X Kinase Buffer R may be shipped frozen or at room temperature. It should be stored at room temperature (20-30°C).

Documents

Manuals & protocols

Recommended products