Kinase Buffer Q (5X)

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


Kinase buffer that has been specially optimized for use with PIK3C3 (hVPS34).
For Research Use Only. Not for use in diagnostic procedures.


Form: Liquid
Product Size: 4 mL
Shipping Condition: Approved for shipment at Room Temperature or on Wet or Dry Ice

Contents & storage

5X Kinase Buffer Q may be shipped frozen or at room temperature. It should be stored at room temperature (20-30°C).


Manuals & protocols

Recommended products