GFP-STAT3

Catalog number: PV5212

 Related applications: Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

STAT3 expressed as a fusion with GFP (Green Fluorescent Protein) to act as a physiological substrate for the Janus family of tyrosine kinases.

For use in combination with the LanthaScreen® Tb-anti-STAT3 [pTyr705] Antibody (PV4846) to evaluate inhibitors in medium to high throughput screening applications.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: Fluorescent
Label or Dye: GFP (EmGFP)
Molecular Weight: 145200 Da
Product Size: 10 nmol
Substrate: STAT3
Substrate Properties: Protein-Based Substrate
Substrate Type: Kinase Substrate
Target Enzyme: Tyrosine Kinases
Validated Kinases: JAK1, JAK2, JAK3, TYK2
Shipping Condition: Dry Ice

Contents & storage

GFP-STAT3 protein substrate in 50 mM Tris-HCl pH 7.5, 100 mM NaCl, 10% Glycerol, 1 mM DTT. Store at -80°C.

Documents

Manuals & protocols

Recommended products