Kinase Tracer 236, 50 µM

Catalog number: PV5592

 Related applications: Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

In the LanthaScreen® Eu Kinase Binding Assay, binding of an Alexa Fluor® conjugate or “tracer" such as Kinase Tracer 236 to a kinase is detected by addition of a Eu-labeled anti-tag antibody. Binding of the tracer & antibody to a kinase results in a high degree of FRET, whereas displacement of the tracer with a kinase inhibitor results in a loss of FRET. Unlike many kinase activity assays, this assay is a simple mix and read assay, with no development steps. Learn More and See Which Kinases Have Been Validated Using This Assay
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format: 384-well plate
Readout: Kinetic
Technique: TR-FRET
Assay Type: Kinase Binding Assay
Label or Dye: Alexa Fluor™ 647
Product Size: 25 µL
Target Enzyme: Kinases (Multiple)
For Use With (Equipment): Microplate Reader
Shipping Condition: Dry Ice

Contents & storage

Store at -20°C.

Documents

Manuals & protocols

Recommended products