LanthaScreen® Cellular Assay Lysis Buffer

Catalog number: PV5598

 Related applications: Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

LanthaScreen® Cellular Assay Lysis buffer is specially formulated for use with the LanthaScreen® Cellular Assays.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reagent Type: Cell Lysis Buffer
Buffer Type: Cell Lysis Buffer
Form: Liquid
Product Line: LanthaScreen®
Product Size: 100 mL
Shipping Condition: Wet Ice

Contents & storage

LanthaScreen® Cellular Assay Lysis buffer is shipped and stored at 4°C.

Documents

Manuals & protocols

Recommended products