LanthaScreen™ Eu-PY20 Antibody Kit

Catalog number:  PV5696

 Related applications: Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

A general reagent to monitor tyrosine kinase activity, the PY20 antibody binds to phosphorylated tyrosines within a peptide or physiological protein substrate. For use in combination with AlexaFluor 647-poly-GT (PV5693) to evaluate inhibitors in medium to high throughput screening applications.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color: Far-Red
Excitation⁄Emission (nm): 340⁄615
Label or Dye: Europium
Product Line: LanthaScreen®
Product Size: 1 mg
Technique: TR-FRET
Validated Kinases: EGFR
Shipping Condition: Dry Ice

Contents & storage

Europium-labeled anti-PY20 antibody in HEPES buffered saline. Store at -20°C.

Documents

Manuals & protocols