Alexa Fluor® 647-Poly-GT Peptide Substrate

Catalog number: PV5836

 Related applications: Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

A general AlexaFluor647-labeled peptide substrate consisting of a polymer of glutamate, and tyrosine that is used to monitor tyrosine kinase acitivty.
For use in combination with the LanthaScreen® Eu-PY20 Antibody (PV5696) to evaluate inhibitors in medium to high throughput screening applications.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color: Far-Red
Detection Method: Fluorescent
Excitation⁄Emission (nm): 340⁄665
Form: Liquid
Label or Dye: Alexa Fluor™ 647
Product Size: 10 nmol
Substrate: Random (Glu:Tyr) polymer in a ratio of 4:1
Substrate Properties: Peptide-Based Substrate
Substrate Type: Kinase Substrate
Target Enzyme: Tyrosine Kinases
Technique: TR-FRET
Validated Kinases: EGFR
Shipping Condition: Dry Ice

Contents & storage

AlexaFluor® 647 Poly-GT peptide substrate in HEPES buffered saline. Store at -20°C.

Documents

Manuals & protocols

Recommended products