LanthaScreen® Elite Tb-anti-HIS Antibody

Catalog number: PV5895

 Related applications: Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

LanthaScreen Elite Terbium-labeled anti-His-tag antibodies are used to indirectly label proteins tagged with 6XHis. This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using amine-reactive Tb chelates negatively impacts protein activity. The Elite Tb-anti-HIS-tag antibody is a specialized antibody with a broader HIS-tag binding performance than our traditional Tb-anti-HIS-tag antibody.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Excitation⁄Emission (nm): 340⁄490, 520
Form: Frozen
Label or Dye: FITC (Fluorescein), Tb (Terbium)
Product Line: LanthaScreen®
Product Size: 1 mg
Shipping Condition: Dry Ice

Contents & storage

Elite Terbium-labeled anti-HIS antibody in HEPES buffered saline. Store at -20°C.

Documents

Manuals & protocols