LanthaScreen® Eu-Streptavidin

Catalog number: PV6025

 Related applications: Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

A general reagent for use in assay development, the Europium-Streptavidin binds to biotinylated peptide or physiological protein substrates.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: Alexa Fluor™ 647, Europium
Form: Frozen
Product Size: 1 mg
Concentration: 0.25 mg⁄ml
Shipping Condition: Dry Ice
Excitation⁄Emission (nm): 340⁄615, 665

Contents & storage

Europium-labeled Streptavidin in HEPES buffered saline. Store at -20°C.

Documents

Manuals & protocols