LanthaScreen® Tb-anti-Histone H3 [AcLys9] Antibody

Catalog number: PV6046

 Related applications: Chromatin Biology | Pharma & Biopharma | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The LanthaScreen® antibody detects the acetylation of Histone H3 at Lys9.

For use in combination with the BacMam Histone H3 [AcLys9] Cellular Assay (A12897) to evaluate inhibitors in medium to high throughput screening applications.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration: 0.25 mg⁄ml
Excitation⁄Emission (nm): 340⁄490, 520
For Use With (Equipment): Microplate Reader
Form: Frozen
High Throughput Compatibility: High Throughput-Compatible
Label or Dye: Tb (Terbium)
Mass: 25 µg
Product Line: LanthaScreen®
Product Size: 25 µg
Target Gene: Histone H3
Target Modification State: Acetylated
Shipping Condition: Dry Ice

Contents & storage

Terbium-labeled Anti-Histone H3 [AcLys9] antibody in HEPES buffered saline. Store at -20°C.

Documents

Manuals & protocols