Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

Qdot™ 800 ITK™ Streptavidin Conjugate Kit

Catalog number:  Q10071MP

Invitrogen™  Related applications: Cellular Imaging | Labeling Chemistry | Protein Labeling & Crosslinking

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Qdot® 800 ITK™ Streptavidin Conjugate Kit provides the Qdot® streptavidin conjugate as 250 µL of a 2 µM solution. Unlike the Qdot® streptavidin conjugates, the Qdot® ITK™ streptavidin conjugates have the streptavidin covalently attached to the inner amphiphilic coating without a PEG linker. These Qdo® conjugates can be used for binding biotinylated probes. The kit also includes 30 mL of Qdot® incubation buffer.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: Qdot® 800
Product Line: ITK™, Qdot®
Product Size: 1 kit
Shipping Condition: Room Temperature

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols