BioProbes® Journal, ProbesOnline™, and BioPath™ E-newsletters. Subscribe today!

Qdot® 565 Streptavidin Conjugate

Catalog number: Q10133MP

Molecular Probes™  Related applications: Cellular Imaging | Labeling Chemistry | Secondary Detection

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Qdot® 565 Streptavidin Conjugate comprises a biotin-binding protein (streptavidin) covalently attached to a fluorescent label (Qdot® nanocrystal). Streptavidin has a very high binding affinity for biotin, and a conjugate of streptavidin is commonly used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids, and other molecules (for example, a biotinylated primary antibody bound to a protein target can be detected with a fluorescently labeled streptavidin). Strategies similar to this are used in many detection protocols including western blots, flow cytometry, imaging and microscopy, and microplate assays, and are also used in purification workflows to achieve targeted fractionation. Qdot® nanocrystal conjugates are provided as 1 μM solutions.

Important Features of Qdot® Streptavidin Conjugates:

• Streptavidin Qdot® 565 conjugate has emission maxima of ~565 nm
• Approximately 5 to 10 streptavidins per Qdot® nanocrystal
• Extremely photostable and bright fluorescence
• Efficiently excited with single-line excitation sources
• Narrow emission, large Stokes shift
• Available in multiple colors
• Ideal for western blots, flow cytometry, imaging and microscopy, microplate assays and more

Properties of Qdot® Nanocrystals
The Qdot® streptavidin conjugate is the size of a large macromolecule or protein (~15–20 nm) and represents the brightest class of streptavidin detection reagents. Qdot® streptavidin conjugates are made from a nanometer-scale crystal of a semiconductor material (CdSe), which is coated with an additional semiconductor shell (ZnS) to improve the optical properties of the material. The Qdot® 705 and Qdot® 800 streptavidin conjugates, which include CdSeTe, are made in a similar fashion. This core-shell material is further coated with a polymer shell that allows the materials to be conjugated to biological molecules and to retain their optical properties.

Other Fluorescent Conjugates of Streptavidin are Available
We offer several other Qdot® colors or try the Qdot® Streptavidin Sampler Kit, containing Qdot® streptavidin conjugates in six colors 525, 565, 585, 605, 655, and 705. In addition to nanocrystal conjugates, we offer a wide range of streptavidins conjugated to Alexa Fluor® dyes, Oregon Green® dye, enzyme conjugates, and traditional fluorophores like Texas Red® dye, fluorescein (FITC) and more.

Find Biotinylated Conjugates
We offer an extensive line of biotinylated conjugates for use in biotin-streptavidin detection strategies.

• Use the primary antibody search tool to find biotinylated primary antibodies
• Use the secondary antibody selector tool to find biotinylated secondary antibodies and biotinylated anti-dye and anti-hapten antibodies

Blocking Endogenous Biotin
Naturally occurring biotins can interfere with biotin-streptavidin detection schemes. For experiments involving fixed and permeabilized cells, try our Endogenous Biotin-Blocking Kit to minimize this interference.

Related Links:
Learn more about Avidin-Biotin Detection
To learn more about Qdot® Nanocrystals

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

Specifications

Label or Dye: Qdot® 565
Product Size: 50 µL
Shipping Condition: Room Temperature

Contents & storage

1 tube — Store at 2 to 6°C; DO NOT FREEZE

Documents

Manuals & protocols

Recommended products